siRNA Details
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197104.1 | NP_001184033.1 | 3 | 583 | ||
NM_005933.3 | NP_005924.2 | 3 | 583 | |||
XM_011542829.2 | XP_011541131.1 | 4 | 674 | |||
XM_011542830.2 | XP_011541132.1 | 4 | 674 | |||
XM_011542831.2 | XP_011541133.1 | 4 | 674 | |||
GenBank mRNA | AF024540.1 | - | 202 | |||
AY187942.1 | - | 239 | ||||
AY187944.1 | - | 368 | ||||
D14540.1 | - | 3 | 583 | |||
L04284.1 | - | 3 | 560 | |||
L04731.1 | - | 3 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
