siRNA Details
Gene Symbol: UCHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270952.1 | NP_001257881.1 | 4, 5 | 482 | ||
NM_006002.4 | NP_005993.1 | 4, 5 | 404 | |||
XM_011535212.1 | XP_011533514.1 | 4, 5 | 404 | |||
XM_011535213.2 | XP_011533515.1 | 3, 4 | 385 | |||
XM_011535214.2 | XP_011533516.1 | 4, 5 | 472 | |||
XM_017020725.1 | XP_016876214.1 | 4, 5 | 404 | |||
XM_017020726.1 | XP_016876215.1 | 4, 5 | 404 | |||
XM_017020727.1 | XP_016876216.1 | 3, 4 | 379 | |||
GenBank mRNA | AK313665.1 | - | 4, 5 | 394 | ||
BC018125.2 | - | 4, 5 | 363 | 3887201 | ||
BQ427804.1 | - | 4, 5 | 418 | |||
BT019359.1 | - | 4, 5 | 333 | |||
CR456855.1 | - | 4, 5 | 333 | |||
DA820150.1 | - | 4, 5 | 404 | |||
M30496.1 | - | 4, 5 | 353 |

More Information
Other Species Targeted

Gene Ontology Categories:
