siRNA Details
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 19 | 2535 | ||
NM_006313.2 | NP_006304.1 | 18 | 2448 | |||
XM_005269261.2 | XP_005269318.1 | 13 | 1739 | |||
XM_017020284.1 | XP_016875773.1 | 17 | 3167 | |||
XM_017020285.1 | XP_016875774.1 | 20 | 3872 | |||
GenBank mRNA | AB011101.2 | - | 18 | 2373 | ||
AF013990.1 | - | 18 | 2331 | |||
AF106069.1 | - | 18 | 2382 | |||
AF153604.1 | - | 19 | 2517 | |||
BC125123.1 | - | 18 | 2382 | 40118994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
