siRNA Details
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 22 | 4534 | ||
NM_006313.2 | NP_006304.1 | 21 | 4447 | |||
XM_005269261.2 | XP_005269318.1 | 16 | 3738 | |||
XM_017020284.1 | XP_016875773.1 | 20 | 5166 | |||
XM_017020285.1 | XP_016875774.1 | 23 | 5871 | |||
GenBank mRNA | AB011101.2 | - | 21 | 4372 | ||
AF106069.1 | - | 21 | 4381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
