siRNA Details
Gene Symbol: USP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252078.1 | NP_001239007.1 | 9, 10 | 1164 | ||
NM_006313.2 | NP_006304.1 | 8, 9 | 1077 | |||
XM_005269261.2 | XP_005269318.1 | 3, 4 | 368 | |||
XM_006719718.3 | XP_006719781.1 | 9, 10 | 1162 | |||
XM_017020284.1 | XP_016875773.1 | 7, 8 | 1796 | |||
XM_017020285.1 | XP_016875774.1 | 10, 11 | 2501 | |||
GenBank mRNA | AB011101.2 | - | 8, 9 | 1002 | ||
AF013990.1 | - | 8, 9 | 960 | |||
AF106069.1 | - | 8, 9 | 1011 | |||
AF153604.1 | - | 9, 10 | 1146 | |||
BC125123.1 | - | 8, 9 | 1011 | 40118994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
