siRNA Details
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276325.1 | NP_001263254.1 | 2 | 563 | ||
NM_006515.3 | NP_006506.3 | 2 | 563 | |||
XM_017007032.1 | XP_016862521.1 | 2 | 626 | |||
GenBank mRNA | AF054989.1 | - | 2 | 489 | ||
AK222734.1 | - | 2 | 499 | |||
AK307537.1 | - | 2 | 563 | |||
AK311418.1 | - | 2 | 596 | |||
AY952295.1 | - | 2 | 468 | |||
BC011635.2 | - | 2 | 489 | 4080032 | ||
U80776.1 | - | 2 | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
