siRNA Details
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243723.1 | NP_001230652.1 | 4 | 1107 | ||
NM_001320676.1 | NP_001307605.1 | 3 | 775 | |||
NM_001320677.1 | NP_001307606.1 | 4 | 1087 | |||
NM_001320678.1 | NP_001307607.1 | 2 | 660 | |||
NM_006515.3 | NP_006506.3 | 3 | 1524 | |||
XM_006713292.3 | XP_006713355.1 | 3 | 860 | |||
XM_006713294.3 | XP_006713357.1 | 4 | 1149 | |||
XM_017007032.1 | XP_016862521.1 | 3 | 1587 | |||
GenBank mRNA | AF054989.1 | - | 3 | 1450 | ||
AK122967.1 | - | 1 | 1720 | |||
AK222734.1 | - | 3 | 1460 | |||
AK297865.1 | - | 4 | 1022 | |||
AK302296.1 | - | 4 | 1041 | |||
AK307537.1 | - | 3 | 1524 | |||
AY952295.1 | - | 3 | 1429 | |||
BC008931.2 | - | 3 | 694 | |||
U80776.1 | - | 3 | 1441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
