siRNA Details
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243723.1 | NP_001230652.1 | 4 | 1523 | ||
NM_001320676.1 | NP_001307605.1 | 3 | 1191 | |||
NM_001320677.1 | NP_001307606.1 | 4 | 1503 | |||
NM_001320678.1 | NP_001307607.1 | 2 | 1076 | |||
NM_006515.3 | NP_006506.3 | 3 | 1940 | |||
XM_006713292.3 | XP_006713355.1 | 3 | 1276 | |||
XM_006713294.3 | XP_006713357.1 | 4 | 1565 | |||
XM_017007032.1 | XP_016862521.1 | 3 | 2003 | |||
GenBank mRNA | AF054989.1 | - | 3 | 1866 | ||
AK122967.1 | - | 1 | 2136 | |||
AK222734.1 | - | 3 | 1876 | |||
AK297865.1 | - | 4 | 1438 | |||
AK302296.1 | - | 4 | 1457 | |||
AK307537.1 | - | 3 | 1940 | |||
AY952295.1 | - | 3 | 1845 | |||
BC008931.2 | - | 3 | 1110 | |||
U80776.1 | - | 3 | 1857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
