siRNA Details
Gene Symbol: PLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252226.1 | NP_001239155.1 | 4, 5 | 640 | ||
NM_006622.3 | NP_006613.2 | 3, 4 | 682 | |||
GenBank mRNA | AF059617.1 | - | 3, 4 | 802 | ||
AF223574.1 | - | 3, 4 | 625 | |||
AK027851.1 | - | 3, 4 | 627 | |||
AK098163.1 | - | 4, 5 | 608 | |||
AK297298.1 | - | 4, 5 | 585 | |||
AK304746.1 | - | 2, 3 | 502 | |||
AK312505.1 | - | 3, 4 | 630 | |||
BC013879.2 | - | 3, 4 | 615 | 3831747 | ||
U85755.1 | - | 2, 3 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
