siRNA Details
Gene Symbol: PLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252226.1 | NP_001239155.1 | 12 | 1634 | ||
NM_006622.3 | NP_006613.2 | 11 | 1676 | |||
GenBank mRNA | AF059617.1 | - | 11 | 1796 | ||
AF223574.1 | - | 11 | 1619 | |||
AK027851.1 | - | 11 | 1620 | |||
AK098163.1 | - | 10 | 1216 | |||
AK312505.1 | - | 11 | 1624 | |||
BC013879.2 | - | 11 | 1609 | 3831747 | ||
U85755.1 | - | 10 | 1171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
