siRNA Details
Gene Symbol: PLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252226.1 | NP_001239155.1 | 14, 15 | 2005 | ||
NM_006622.3 | NP_006613.2 | 13, 14 | 2047 | |||
GenBank mRNA | AF059617.1 | - | 13, 14 | 2167 | ||
AF223574.1 | - | 13, 14 | 1990 | |||
AK027851.1 | - | 13, 14 | 1991 | |||
AK098163.1 | - | 12, 13 | 1588 | |||
AK312505.1 | - | 13, 14 | 1995 | |||
BC013879.2 | - | 13, 14 | 1980 | 3831747 | ||
U85755.1 | - | 12, 13 | 1542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
