siRNA Details
Gene Symbol: RUVBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321190.1 | NP_001308119.1 | 14 | 1865 | ||
NM_001321191.1 | NP_001308120.1 | 14 | 1774 | |||
NM_006666.2 | NP_006657.1 | 14 | 1795 | |||
NR_135578.1 | 14 | 1784 | ||||
XM_011526330.1 | XP_011524632.1 | 14 | 1708 | |||
GenBank mRNA | AB024301.1 | - | 14 | 1344 | ||
AF124607.1 | - | 14 | 1352 | |||
AF151804.1 | - | 14 | 1336 | |||
AF155138.1 | - | 14 | 1337 | |||
AK027762.1 | - | 14 | 1415 | |||
AK057498.1 | - | 14 | 2342 | |||
AK074542.1 | - | 14 | 1345 | |||
AL136743.1 | - | 13 | 1334 | |||
BC000428.2 | - | 14 | 1336 | 2819778 | ||
BC004531.2 | - | 14 | 1344 | 3051517 | ||
BC008355.1 | - | 14 | 1333 | |||
CR533507.1 | - | 13 | 1331 | |||
Y18417.1 | - | 14 | 1336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
