siRNA Details
Gene Symbol: RUVBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321190.1 | NP_001308119.1 | 14 | 1803 | ||
NM_001321191.1 | NP_001308120.1 | 14 | 1712 | |||
NM_006666.2 | NP_006657.1 | 14 | 1733 | |||
NR_135578.1 | 14 | 1722 | ||||
XM_011526330.1 | XP_011524632.1 | 14 | 1646 | |||
GenBank mRNA | AB024301.1 | - | 14 | 1282 | ||
AF124607.1 | - | 14 | 1290 | |||
AF151804.1 | - | 14 | 1274 | |||
AF155138.1 | - | 14 | 1275 | |||
AK027762.1 | - | 14 | 1353 | |||
AK057498.1 | - | 14 | 2280 | |||
AK074542.1 | - | 14 | 1283 | |||
AL136743.1 | - | 13 | 1272 | |||
BC000428.2 | - | 14 | 1274 | 2819778 | ||
BC004531.2 | - | 14 | 1282 | 3051517 | ||
BC008355.1 | - | 14 | 1271 | |||
CR533507.1 | - | 13 | 1269 | |||
Y18417.1 | - | 14 | 1274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
