siRNA Details
Gene Symbol: COPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006837.2 | NP_006828.2 | 8 | 1279 | ||
GenBank mRNA | AB209134.1 | - | 6 | 5483 | ||
AK056967.1 | - | 10 | 1055 | |||
AK128519.1 | - | 7 | 3271 | |||
BC001187.1 | - | 8 | 1083 | 3354130 | ||
BC001859.2 | - | 8 | 1061 | 3530890 | ||
BC007272.1 | - | 8 | 1057 | 3161025 | ||
BX648542.1 | - | 8 | 3641 | |||
CR541678.1 | - | 8 | 948 | |||
U65928.1 | - | 8 | 1075 | |||
U70734.1 | - | 8 | 1068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
