siRNA Details
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012238.4 | NP_036370.2 | 2, 3 | 602 | ||
GenBank mRNA | AF083106.2 | - | 2, 3 | 602 | ||
AF235040.1 | - | 2, 3 | 597 | |||
AK289743.1 | - | 2, 3 | 338 | |||
AL136741.1 | - | 2, 3 | 217 | |||
BC012499.1 | - | 2, 3 | 182 | 4518906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
