siRNA Details
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142498.1 | NP_001135970.1 | 5 | 660 | ||
NM_001314049.1 | NP_001300978.1 | 4 | 449 | |||
NM_012238.4 | NP_036370.2 | 6 | 1166 | |||
GenBank mRNA | AF083106.2 | - | 6 | 1166 | ||
AF235040.1 | - | 6 | 1161 | |||
AK027686.1 | - | 3 | 223 | |||
AK074805.1 | - | 4 | 367 | |||
AK289743.1 | - | 6 | 902 | |||
AL136741.1 | - | 6 | 781 | |||
BC012499.1 | - | 6 | 746 | 4518906 | ||
BX648554.1 | - | 3 | 409 | |||
JQ768366.1 | - | 4 | 537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
