siRNA Details
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 3 | 490 | ||
NM_012239.5 | NP_036371.1 | 3 | 636 | |||
XM_005252835.1 | XP_005252892.1 | 3 | 636 | |||
XM_011519956.1 | XP_011518258.1 | 3 | 1428 | |||
XM_011519957.1 | XP_011518259.1 | 3 | 1257 | |||
XM_017017428.1 | XP_016872917.1 | 3 | 482 | |||
XM_017017429.1 | XP_016872918.1 | 3 | 656 | |||
XM_017017430.1 | XP_016872919.1 | 3 | 650 | |||
XM_017017431.1 | XP_016872920.1 | 3 | 381 | |||
GenBank mRNA | AF083108.2 | - | 3 | 704 | ||
AK299009.1 | - | 3 | 626 | |||
AK299438.1 | - | 3 | 468 | |||
AK301558.1 | - | 2 | 436 | |||
AK301979.1 | - | 3 | 624 | |||
AK308681.1 | - | 3 | 637 | |||
AK316175.1 | - | 3 | 540 | |||
AL535769.3 | - | 3 | 471 | |||
BC001042.2 | - | 3 | 619 | 2823471 | ||
BI755839.1 | - | 3 | 436 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
