siRNA Details
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 5 | 760 | ||
NM_012239.5 | NP_036371.1 | 5 | 906 | |||
XM_005252835.1 | XP_005252892.1 | 5 | 906 | |||
XM_011519956.1 | XP_011518258.1 | 5 | 1698 | |||
XM_011519957.1 | XP_011518259.1 | 5 | 1527 | |||
XM_017017428.1 | XP_016872917.1 | 5 | 752 | |||
XM_017017429.1 | XP_016872918.1 | 5 | 926 | |||
XM_017017430.1 | XP_016872919.1 | 5 | 920 | |||
XM_017017431.1 | XP_016872920.1 | 5 | 651 | |||
GenBank mRNA | AF083108.2 | - | 5 | 974 | ||
AK074992.1 | - | 2 | 209 | |||
AK298572.1 | - | 5 | 655 | |||
AK299438.1 | - | 5 | 738 | |||
AK301558.1 | - | 4 | 706 | |||
AK301979.1 | - | 5 | 894 | |||
AK316175.1 | - | 5 | 810 | |||
AL137276.1 | - | 1 | 29 | |||
AL535769.3 | - | 5 | 741 | |||
BC001042.2 | - | 5 | 889 | 2823471 | ||
BI755839.1 | - | 5 | 706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
