siRNA Details
Gene Symbol: SIRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017524.2 | NP_001017524.1 | 7 | 1626 | ||
NM_012239.5 | NP_036371.1 | 7 | 1772 | |||
XM_005252835.1 | XP_005252892.1 | 7 | 1826 | |||
XM_011519956.1 | XP_011518258.1 | 7 | 2618 | |||
XM_011519957.1 | XP_011518259.1 | 7 | 2447 | |||
XM_017017428.1 | XP_016872917.1 | 7 | 1672 | |||
XM_017017429.1 | XP_016872918.1 | 7 | 1846 | |||
XM_017017430.1 | XP_016872919.1 | 7 | 1786 | |||
XM_017017431.1 | XP_016872920.1 | 7 | 1571 | |||
GenBank mRNA | AF083108.2 | - | 7 | 1840 | ||
AK074992.1 | - | 5 | 2612 | |||
AK301979.1 | - | 7 | 1814 | |||
AL137276.1 | - | 3 | 895 | |||
BC001042.2 | - | 7 | 1755 | 2823471 | ||
DR762907.1 | - | 1 | 486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
