siRNA Details
Gene Symbol: PRDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012406.3 | NP_036538.3 | 9 | 1930 | ||
XM_005268593.1 | XP_005268650.1 | 9 | 1949 | |||
GenBank mRNA | AF144757.2 | - | 8 | 1615 | ||
AK056378.1 | - | 9 | 1797 | |||
AL133083.1 | - | 6 | 1265 | |||
BC035581.1 | - | 8 | 1731 | 5587364 | ||
BC068562.1 | - | 9 | 1631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
