siRNA Details
Gene Symbol: PLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB006972.1 | - | 13 | 2679 | ||
AK302858.1 | - | 13 | 2588 | |||
AK314238.1 | - | 13 | 2733 | |||
Y13115.1 | - | 13 | 2631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
