siRNA Details
Gene Symbol: PLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190799.1 | NP_001177728.1 | 14 | 2977 | ||
NM_001190801.1 | NP_001177730.1 | 15 | 2896 | |||
NM_014264.4 | NP_055079.3 | 15 | 3073 | |||
XM_005262701.2 | XP_005262758.1 | 15 | 2933 | |||
XM_017007662.1 | XP_016863151.1 | 15 | 3232 | |||
XM_017007663.1 | XP_016863152.1 | 15 | 3131 | |||
GenBank mRNA | AB006972.1 | - | 15 | 2987 | ||
AK302858.1 | - | 15 | 2896 | |||
AK303399.1 | - | 14 | 2911 | |||
AK314238.1 | - | 15 | 3041 | |||
BC036023.1 | - | 15 | 3041 | 5273226 | ||
Y13115.1 | - | 15 | 2939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
