siRNA Details
Gene Symbol: KMT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014727.2 | NP_055542.1 | 3 | 2403 | ||
XM_011527561.2 | XP_011525863.2 | 3 | 2274 | |||
XM_011527562.2 | XP_011525864.1 | 3 | 2604 | |||
XM_017027544.1 | XP_016883033.1 | 3 | 2604 | |||
XM_017027545.1 | XP_016883034.1 | 2 | 2317 | |||
GenBank mRNA | AB002302.3 | - | 1 | 1503 | ||
AF104918.1 | - | 1 | 417 | |||
AF186605.1 | - | 3 | 2075 | |||
AJ007041.1 | - | 3 | 2403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
