siRNA Details
Gene Symbol: KMT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014727.2 | NP_055542.1 | 28 | 6564 | ||
XM_011527561.2 | XP_011525863.2 | 28 | 6435 | |||
XM_011527562.2 | XP_011525864.1 | 28 | 6765 | |||
XM_017027544.1 | XP_016883033.1 | 28 | 6765 | |||
XM_017027545.1 | XP_016883034.1 | 27 | 6478 | |||
XM_017027546.1 | XP_016883035.1 | 23 | 3686 | |||
GenBank mRNA | AB002302.3 | - | 26 | 5664 | ||
AF186605.1 | - | 28 | 6236 | |||
AJ007041.1 | - | 28 | 6564 | |||
BC009337.2 | - | 1 | 804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
