siRNA Details
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 5 | 733 | ||
NM_014903.5 | NP_055718.4 | 5 | 733 | |||
XM_005269215.3 | XP_005269272.1 | 5 | 4734 | |||
XM_011538940.2 | XP_011537242.1 | 5 | 4735 | |||
XM_011538941.2 | XP_011537243.1 | 5 | 4734 | |||
XM_011538944.2 | XP_011537246.1 | 5 | 4732 | |||
XM_017020164.1 | XP_016875653.1 | 5 | 4736 | |||
XM_017020165.1 | XP_016875654.1 | 5 | 4736 | |||
XM_017020166.1 | XP_016875655.1 | 5 | 722 | |||
XM_017020167.1 | XP_016875656.1 | 5 | 722 | |||
XM_017020168.1 | XP_016875657.1 | 5 | 4735 | |||
XM_017020169.1 | XP_016875658.1 | 5 | 4734 | |||
XM_017020170.1 | XP_016875659.1 | 5 | 4731 | |||
GenBank mRNA | AF397731.1 | - | 4 | 317 | ||
AJ488201.1 | - | 5 | 605 | |||
BC017667.2 | - | 5 | 1110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
