siRNA Details
Gene Symbol: NAV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024383.1 | NP_001019554.1 | 16 | 4296 | ||
NM_014903.5 | NP_055718.4 | 16 | 4296 | |||
XM_005269215.3 | XP_005269272.1 | 16 | 8297 | |||
XM_011538940.2 | XP_011537242.1 | 16 | 8298 | |||
XM_011538941.2 | XP_011537243.1 | 16 | 8297 | |||
XM_017020164.1 | XP_016875653.1 | 16 | 8299 | |||
XM_017020165.1 | XP_016875654.1 | 16 | 8299 | |||
XM_017020166.1 | XP_016875655.1 | 16 | 4285 | |||
XM_017020167.1 | XP_016875656.1 | 15 | 4216 | |||
XM_017020171.1 | XP_016875660.1 | 8 | 7592 | |||
XM_017020172.1 | XP_016875661.1 | 8 | 3230 | |||
GenBank mRNA | AF397731.1 | - | 15 | 3880 | ||
AJ488201.1 | - | 16 | 4168 | |||
AK298525.1 | - | 1 | 52 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
