siRNA Details
Gene Symbol: HDAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098416.3 | NP_001091886.1 | 25 | 3051 | ||
NM_001308090.1 | NP_001295019.1 | 25 | 3111 | |||
NM_015401.4 | NP_056216.2 | 26 | 3162 | |||
XM_011538478.1 | XP_011536780.1 | 26 | 3114 | |||
XM_011538479.1 | XP_011536781.1 | 27 | 3556 | |||
XM_011538480.1 | XP_011536782.1 | 25 | 3003 | |||
XM_011538481.1 | XP_011536783.1 | 26 | 3007 | |||
XM_011538482.1 | XP_011536784.1 | 26 | 3091 | |||
XM_017019455.1 | XP_016874944.1 | 25 | 3063 | |||
GenBank mRNA | AF239243.1 | - | 23 | 2955 | ||
AK001032.1 | - | 10 | 902 | |||
AK001190.1 | - | 18 | 2084 | |||
AK024387.1 | - | 3896 | ||||
AK024469.1 | - | 21 | 3142 | |||
AK026767.1 | - | 2 | 917 | |||
AK027781.1 | - | 8 | 801 | |||
AK122588.1 | - | 11 | 5290 | |||
AK128383.1 | - | 18 | 2005 | |||
AK299292.1 | - | 26 | 3077 | |||
AK301545.1 | - | 27 | 3132 | |||
AK303481.1 | - | 25 | 3017 | |||
AL117455.1 | - | 22 | 2629 | |||
AY321367.1 | - | 25 | 2936 | |||
BC006453.1 | - | 15 | 1586 | |||
BC020505.2 | - | 18 | 2129 | |||
BC064840.1 | - | 25 | 2945 | 6179239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
