siRNA Details
Gene Symbol: HDAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098416.3 | NP_001091886.1 | 25 | 3229 | ||
NM_001308090.1 | NP_001295019.1 | 25 | 3289 | |||
NM_015401.4 | NP_056216.2 | 26 | 3340 | |||
XM_011538478.1 | XP_011536780.1 | 26 | 3292 | |||
XM_011538479.1 | XP_011536781.1 | 27 | 3734 | |||
XM_011538480.1 | XP_011536782.1 | 25 | 3181 | |||
XM_011538481.1 | XP_011536783.1 | 26 | 3185 | |||
XM_011538482.1 | XP_011536784.1 | 26 | 3269 | |||
XM_017019455.1 | XP_016874944.1 | 25 | 3241 | |||
GenBank mRNA | AK001032.1 | - | 10 | 1080 | ||
AK001190.1 | - | 18 | 2262 | |||
AK024387.1 | - | 4074 | ||||
AK024469.1 | - | 21 | 3320 | |||
AK026767.1 | - | 2 | 1095 | |||
AK027781.1 | - | 8 | 979 | |||
AK122588.1 | - | 11 | 5468 | |||
AK128383.1 | - | 18 | 2183 | |||
AK303481.1 | - | 25 | 3195 | |||
AL117455.1 | - | 22 | 2807 | |||
AY321367.1 | - | 25 | 3114 | |||
BC006453.1 | - | 15 | 1764 | |||
BC020505.2 | - | 18 | 2307 | |||
BC064840.1 | - | 25 | 3123 | 6179239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
