siRNA Details
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278185.1 | NP_001265114.1 | 5 | 742 | ||
NM_001278186.1 | NP_001265115.1 | 4 | 582 | |||
NM_015601.3 | NP_056416.2 | 5 | 742 | |||
NM_022079.2 | NP_071362.1 | 5 | 742 | |||
XM_011539592.2 | XP_011537894.1 | 6 | 816 | |||
XM_011539593.2 | XP_011537895.1 | 6 | 816 | |||
XM_011539594.2 | XP_011537896.1 | 7 | 902 | |||
XM_011539595.2 | XP_011537897.1 | 7 | 952 | |||
XM_011539596.2 | XP_011537898.1 | 6 | 830 | |||
XM_011539597.2 | XP_011537899.1 | 6 | 816 | |||
XM_017016041.1 | XP_016871530.1 | 5 | 744 | |||
GenBank mRNA | AB046813.1 | - | 2 | 596 | ||
AK292809.1 | - | 5 | 622 | |||
AY221963.1 | - | 5 | 679 | |||
AY650032.1 | - | 4 | 519 | |||
AY650033.1 | - | 5 | 679 | |||
BC015043.1 | - | 4 | 485 | |||
BC039600.1 | - | 5 | 652 | 5212048 | ||
BX537563.1 | - | 3 | 420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
