siRNA Details
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278185.1 | NP_001265114.1 | 8 | 1171 | ||
NM_001278186.1 | NP_001265115.1 | 6 | 919 | |||
NM_015601.3 | NP_056416.2 | 8 | 1171 | |||
NM_022079.2 | NP_071362.1 | 8 | 1171 | |||
XM_011539592.2 | XP_011537894.1 | 9 | 1245 | |||
XM_011539593.2 | XP_011537895.1 | 9 | 1245 | |||
XM_011539594.2 | XP_011537896.1 | 10 | 1331 | |||
XM_011539595.2 | XP_011537897.1 | 10 | 1381 | |||
XM_011539596.2 | XP_011537898.1 | 9 | 1259 | |||
XM_011539597.2 | XP_011537899.1 | 9 | 1245 | |||
XM_017016041.1 | XP_016871530.1 | 8 | 1173 | |||
GenBank mRNA | AB046813.1 | - | 5 | 1025 | ||
AK292809.1 | - | 8 | 1051 | |||
AY221963.1 | - | 8 | 1108 | |||
AY650032.1 | - | 6 | 856 | |||
AY650033.1 | - | 8 | 1108 | |||
BC015043.1 | - | 6 | 822 | |||
BC039600.1 | - | 8 | 1081 | 5212048 | ||
BX537563.1 | - | 6 | 849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
