siRNA Details
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278185.1 | NP_001265114.1 | 19, 20 | 2527 | ||
NM_001278186.1 | NP_001265115.1 | 16, 17 | 2251 | |||
NM_015601.3 | NP_056416.2 | 18, 19 | 2503 | |||
NM_022079.2 | NP_071362.1 | 19, 20 | 2527 | |||
XM_011539592.2 | XP_011537894.1 | 20, 21 | 2601 | |||
XM_011539593.2 | XP_011537895.1 | 19, 20 | 2577 | |||
XM_011539594.2 | XP_011537896.1 | 21, 22 | 2687 | |||
XM_011539595.2 | XP_011537897.1 | 21, 22 | 2737 | |||
XM_011539596.2 | XP_011537898.1 | 20, 21 | 2615 | |||
XM_011539597.2 | XP_011537899.1 | 18, 19 | 2238 | |||
XM_017016041.1 | XP_016871530.1 | 17, 18 | 2166 | |||
GenBank mRNA | AB046813.1 | - | 16, 17 | 2381 | ||
AK027835.1 | - | 9, 10 | 1070 | |||
AK292809.1 | - | 18, 19 | 2383 | |||
AL096715.1 | - | 1, 2 | 1900 | |||
AY221963.1 | - | 19, 20 | 2464 | |||
AY650032.1 | - | 16, 17 | 2188 | |||
AY650033.1 | - | 19, 20 | 2464 | |||
BC015043.1 | - | 16, 17 | 2154 | |||
BC039600.1 | - | 18, 19 | 2413 | 5212048 | ||
BX537563.1 | - | 18, 19 | 4990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
