siRNA Details
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320687.1 | NP_001307616.1 | 7 | 1549 | ||
NM_016166.2 | NP_057250.1 | 6 | 950 | |||
XM_011522126.2 | XP_011520428.1 | 8 | 1657 | |||
XM_011522127.2 | XP_011520429.1 | 6 | 1434 | |||
XM_017022688.1 | XP_016878177.1 | 6 | 1428 | |||
XM_017022689.1 | XP_016878178.1 | 6 | 1441 | |||
XM_017022690.1 | XP_016878179.1 | 6 | 1246 | |||
XM_017022691.1 | XP_016878180.1 | 6 | 1020 | |||
GenBank mRNA | AF167160.1 | - | 6 | 898 | ||
AK094641.1 | - | 7 | 1549 | |||
AK314515.1 | - | 6 | 825 | |||
AY826819.1 | - | 6 | 799 | |||
BC053685.1 | - | 6 | 803 | |||
BC104646.1 | - | 6 | 818 | |||
BC118587.1 | - | 6 | 852 | 40085010 | ||
BC121797.1 | - | 6 | 849 | 40085007 | ||
EF487997.1 | - | 6 | 802 | |||
EF487998.1 | - | 6 | 802 | |||
FJ997900.1 | - | 6 | 802 | |||
U78524.1 | - | 5 | 784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
