siRNA Details
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320687.1 | NP_001307616.1 | 3, 4 | 1212 | ||
NM_016166.2 | NP_057250.1 | 2, 3 | 613 | |||
XM_011522126.2 | XP_011520428.1 | 4, 5 | 1320 | |||
XM_011522127.2 | XP_011520429.1 | 2, 3 | 1097 | |||
XM_017022688.1 | XP_016878177.1 | 2, 3 | 1091 | |||
XM_017022689.1 | XP_016878178.1 | 2, 3 | 1104 | |||
XM_017022690.1 | XP_016878179.1 | 2, 3 | 909 | |||
XM_017022691.1 | XP_016878180.1 | 2, 3 | 683 | |||
GenBank mRNA | AF077951.1 | - | 2, 3 | 465 | ||
AF167160.1 | - | 2, 3 | 561 | |||
AK094641.1 | - | 3, 4 | 1212 | |||
AK314515.1 | - | 2, 3 | 488 | |||
AY826819.1 | - | 2, 3 | 462 | |||
BC053685.1 | - | 2, 3 | 466 | |||
BC104646.1 | - | 2, 3 | 481 | |||
BC118587.1 | - | 2, 3 | 515 | 40085010 | ||
BC121797.1 | - | 2, 3 | 512 | 40085007 | ||
EF487997.1 | - | 2, 3 | 465 | |||
EF487998.1 | - | 2, 3 | 465 | |||
FJ997900.1 | - | 2, 3 | 465 | |||
U78524.1 | - | 1, 2 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
