siRNA Details
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193285.2 | NP_001180214.1 | 4, 5 | 502 | ||
NM_001321058.1 | NP_001307987.1 | 3, 4 | 374 | |||
NM_001321059.1 | NP_001307988.1 | 3, 4 | 319 | |||
NM_001321060.1 | NP_001307989.1 | 4, 5 | 502 | |||
NM_001321061.1 | NP_001307990.1 | 3, 4 | 374 | |||
NM_001321062.1 | NP_001307991.1 | 2, 3 | 191 | |||
NM_001321063.1 | NP_001307992.1 | 3, 4 | 319 | |||
NM_001321064.1 | NP_001307993.1 | 4, 5 | 467 | |||
NM_016539.3 | NP_057623.2 | 4, 5 | 502 | |||
GenBank mRNA | AF233396.1 | - | 4, 5 | 497 | ||
AK074810.1 | - | 4, 5 | 465 | |||
AK225567.1 | - | 4, 5 | 470 | |||
AK293348.1 | - | 3, 4 | 335 | |||
AK299420.1 | - | 3, 4 | 317 | |||
AK301247.1 | - | 3, 4 | 282 | |||
AK315048.1 | - | 4, 5 | 466 | |||
AL559383.3 | - | 4, 5 | 440 | |||
BC004218.2 | - | 3, 4 | 314 | 3352203 | ||
BC005026.1 | - | 4, 5 | 440 | 2820472 | ||
BC028220.1 | - | 4, 5 | 448 | 5217599 | ||
BE281103.1 | - | 3, 4 | 300 | |||
BP418952.1 | - | 2, 3 | 136 | |||
BQ060703.1 | - | 3, 4 | 215 | |||
CN364175.1 | - | 4, 5 | 445 | |||
CN364177.1 | - | 4, 5 | 505 | |||
CR457200.1 | - | 4, 5 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
