siRNA Details
Gene Symbol: KMT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300908.1 | NP_001287837.1 | 4, 5 | 749 | ||
NM_016028.4 | NP_057112.3 | 4, 5 | 633 | |||
NM_017635.4 | NP_060105.3 | 4, 5 | 661 | |||
XM_005274035.3 | XP_005274092.2 | 4, 5 | 881 | |||
XM_011545091.1 | XP_011543393.1 | 3, 4 | 455 | |||
XM_011545092.2 | XP_011543394.1 | 3, 4 | 645 | |||
XM_011545093.2 | XP_011543395.1 | 4, 5 | 881 | |||
XM_011545094.2 | XP_011543396.1 | 4, 5 | 881 | |||
XM_017017880.1 | XP_016873369.1 | 4, 5 | 881 | |||
GenBank mRNA | AF264782.1 | - | 3, 4 | 553 | ||
AK025183.1 | - | 3, 4 | 321 | |||
AK314336.1 | - | 4, 5 | 576 | |||
BC002522.2 | - | 4, 5 | 580 | |||
BC065287.1 | - | 4, 5 | 585 | |||
BC087834.1 | - | 4, 5 | 602 | |||
BC098121.1 | - | 3, 4 | 382 | 40001705 | ||
BC099714.1 | - | 3, 4 | 382 | 40001707 | ||
BC103498.1 | - | 3, 4 | 382 | 40001706 | ||
BC104483.1 | - | 3, 4 | 382 | 40001704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
