siRNA Details
Gene Symbol: OTUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017670.2 | NP_060140.2 | 6 | 1176 | ||
NR_003089.1 | 5 | 1384 | ||||
GenBank mRNA | AF161381.1 | - | 6 | 584 | ||
AK000120.1 | - | 6 | 616 | |||
AK098029.1 | - | 5 | 1384 | |||
AK298287.1 | - | 7 | 895 | |||
AK303228.1 | - | 5 | 826 | |||
AK307413.1 | - | 6 | 711 | |||
AL096714.1 | - | 5 | 795 | |||
AY177200.1 | - | 6 | 572 | |||
BC007519.2 | - | 6 | 580 | 3051266 | ||
BC010368.1 | - | 6 | 602 | 4334279 | ||
BC107701.1 | - | 6 | 602 | 6494629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
