siRNA Details
Gene Symbol: UBE2R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017811.3 | NP_060281.2 | 5 | 1894 | ||
XM_005251496.2 | XP_005251553.1 | 4 | 1827 | |||
XM_011517949.1 | XP_011516251.1 | 4 | 1779 | |||
XM_017014860.1 | XP_016870349.1 | 3 | 1692 | |||
GenBank mRNA | AK000426.1 | - | 5 | 1511 | ||
AL133827.1 | - | 1 | 19 | |||
BC047584.1 | - | 5 | 1749 | 4827644 | ||
CN427315.1 | - | 1 | 75 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
