siRNA Details
Gene Symbol: UBE2R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017811.3 | NP_060281.2 | 5 | 1384 | ||
XM_005251496.2 | XP_005251553.1 | 4 | 1317 | |||
XM_011517949.1 | XP_011516251.1 | 4 | 1269 | |||
XM_017014860.1 | XP_016870349.1 | 3 | 1182 | |||
GenBank mRNA | AJ240087.1 | - | 5 | 815 | ||
AK000426.1 | - | 5 | 1001 | |||
AK225880.1 | - | 5 | 989 | |||
BC004862.2 | - | 5 | 991 | 3838157 | ||
BC047584.1 | - | 5 | 1240 | 4827644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
