siRNA Details
Gene Symbol: USP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282659.1 | NP_001269588.1 | 9 | 1747 | ||
NM_017944.3 | NP_060414.3 | 8 | 1543 | |||
XM_005252997.1 | XP_005253054.1 | 10 | 1807 | |||
XM_011520196.1 | XP_011518498.1 | 2 | 922 | |||
XM_017017953.1 | XP_016873442.1 | 10 | 1863 | |||
XM_017017954.1 | XP_016873443.1 | 2 | 2070 | |||
GenBank mRNA | AK027283.1 | - | 655 | |||
AK027362.1 | - | 8 | 948 | |||
AK092290.1 | - | 10 | 1164 | |||
AK127264.1 | - | 9 | 1543 | |||
AK309240.1 | - | 9 | 1211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
