siRNA Details
Gene Symbol: USP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282659.1 | NP_001269588.1 | 9, 10 | 1836 | ||
NM_017944.3 | NP_060414.3 | 8, 9 | 1632 | |||
XM_005252997.1 | XP_005253054.1 | 10, 11 | 1896 | |||
XM_011520196.1 | XP_011518498.1 | 2, 3 | 1011 | |||
XM_017017953.1 | XP_016873442.1 | 10, 11 | 1952 | |||
XM_017017954.1 | XP_016873443.1 | 2, 3 | 2159 | |||
GenBank mRNA | AK027283.1 | - | 744 | |||
AK027362.1 | - | 8, 9 | 1037 | |||
AK092290.1 | - | 10, 11 | 1253 | |||
AK127264.1 | - | 9, 10 | 1632 | |||
AK309240.1 | - | 9, 10 | 1300 | |||
DA490237.1 | - | 1, 2 | 53 |

More Information
Other Species Targeted

Gene Ontology Categories:
