siRNA Details
Gene Symbol: USP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282659.1 | NP_001269588.1 | 17 | 2814 | ||
NM_017944.3 | NP_060414.3 | 16 | 2610 | |||
XM_005252997.1 | XP_005253054.1 | 18 | 2874 | |||
XM_011520196.1 | XP_011518498.1 | 10 | 1989 | |||
XM_017017953.1 | XP_016873442.1 | 18 | 2930 | |||
XM_017017954.1 | XP_016873443.1 | 10 | 3137 | |||
GenBank mRNA | AK027362.1 | - | 16 | 2015 | ||
AK074076.1 | - | 2 | 302 | |||
AK092290.1 | - | 18 | 2230 | |||
AK127264.1 | - | 17 | 2610 | |||
AL832991.2 | - | 2 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
