siRNA Details
Gene Symbol: HDAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166418.1 | NP_001159890.1 | 6, 7 | 987 | ||
NM_018486.2 | NP_060956.1 | 8, 9 | 1260 | |||
NR_051952.1 | 7, 8 | 1118 | ||||
XM_011530986.2 | XP_011529288.2 | 8, 9 | 1255 | |||
XM_011530987.2 | XP_011529289.2 | 8, 9 | 1255 | |||
XM_017029640.1 | XP_016885129.1 | 7, 8 | 1177 | |||
XM_017029641.1 | XP_016885130.1 | 7, 8 | 1177 | |||
XM_017029642.1 | XP_016885131.1 | 6, 7 | 980 | |||
XM_017029643.1 | XP_016885132.1 | 5, 6 | 955 | |||
XM_017029644.1 | XP_016885133.1 | 5, 6 | 904 | |||
XM_017029645.1 | XP_016885134.1 | 5, 6 | 955 | |||
XM_017029646.1 | XP_016885135.1 | 7, 8 | 804 | |||
GenBank mRNA | AF230097.1 | - | 8, 9 | 973 | ||
AF245664.1 | - | 8, 9 | 927 | |||
AJ277724.1 | - | 8, 9 | 960 | |||
AK296641.1 | - | 6, 7 | 937 | |||
AK298767.1 | - | 7, 8 | 858 | |||
BC050433.1 | - | 8, 9 | 951 | 5761745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
