siRNA Details
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278945.1 | NP_001265874.1 | 7 | 1104 | ||
NM_018492.3 | NP_060962.2 | 7 | 1129 | |||
XM_006716368.2 | XP_006716431.1 | 7 | 1053 | |||
GenBank mRNA | AB027249.1 | - | 7 | 855 | ||
AB027250.1 | - | 7 | 835 | |||
AF189722.1 | - | 7 | 819 | |||
AF237709.1 | - | 7 | 843 | |||
AK027291.1 | - | 7 | 834 | |||
AK301836.1 | - | 7 | 1064 | |||
BC015191.1 | - | 7 | 843 | 4082846 | ||
EU794685.1 | - | 7 | 866 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
