siRNA Details
Gene Symbol: PBK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278945.1 | NP_001265874.1 | 6 | 913 | ||
NM_018492.3 | NP_060962.2 | 6 | 971 | |||
XM_006716368.2 | XP_006716431.1 | 6 | 895 | |||
GenBank mRNA | AB027249.1 | - | 6 | 697 | ||
AB027250.1 | - | 6 | 677 | |||
AF189722.1 | - | 6 | 661 | |||
AF237709.1 | - | 6 | 685 | |||
AK301836.1 | - | 6 | 873 | |||
BC015191.1 | - | 6 | 685 | 4082846 | ||
EU794685.1 | - | 6 | 708 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023209, BC020099, AK076121, AK003838, AB041882, and others.
Gene Ontology Categories:
