siRNA Details
Gene Symbol: SMYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020197.2 | NP_064582.2 | 12 | 1336 | ||
GenBank mRNA | AF226053.1 | - | 12 | 1325 | ||
AK091590.1 | - | 2 | 3294 | |||
AK098683.1 | - | 8 | 820 | |||
BC017080.2 | - | 5 | 599 | |||
BC028104.1 | - | 11 | 1257 | |||
BC068511.1 | - | 9 | 3342 | |||
BC098133.1 | - | 12 | 1325 | 40005281 | ||
BC098276.1 | - | 12 | 1325 | 40005283 | ||
BC098305.1 | - | 10 | 1029 | 40005284 | ||
BC098335.1 | - | 12 | 1325 | 40005280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
