siRNA Details
Gene Symbol: PRDM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020228.2 | NP_064613.2 | 10, 11 | 1527 | ||
NM_199437.1 | NP_955469.1 | 10, 11 | 1527 | |||
NM_199438.1 | NP_955470.1 | 6, 7 | 1253 | |||
NM_199439.1 | NP_955471.1 | 6, 7 | 1253 | |||
XM_011542912.2 | XP_011541214.1 | 10, 11 | 1789 | |||
XM_017018023.1 | XP_016873512.1 | 6, 7 | 3338 | |||
GenBank mRNA | AB033057.2 | - | 6, 7 | 1037 | ||
AF275817.1 | - | 6, 7 | 1253 | |||
AF503171.1 | - | 6, 7 | 1184 | |||
AK000234.1 | - | 4, 5 | 494 | |||
AK310354.1 | - | 11, 12 | 1573 | |||
AL833172.1 | - | 10, 11 | 1530 | |||
BC112934.1 | - | 10, 11 | 1519 | 6502888 | ||
BC117415.1 | - | 6, 7 | 1092 | 40125966 | ||
BC143612.1 | - | 6, 7 | 1092 | 9052125 | ||
BC143613.1 | - | 6, 7 | 1092 | 9052126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
