siRNA Details
Gene Symbol: PRDM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020228.2 | NP_064613.2 | 22 | 5363 | ||
NM_199437.1 | NP_955469.1 | 21 | 5351 | |||
NM_199438.1 | NP_955470.1 | 18 | 5050 | |||
NM_199439.1 | NP_955471.1 | 16 | 4936 | |||
XM_011542912.2 | XP_011541214.1 | 21 | 5574 | |||
XM_017018023.1 | XP_016873512.1 | 17 | 7123 | |||
GenBank mRNA | AB033057.2 | - | 18 | 4834 | ||
AF275817.1 | - | 18 | 5050 | |||
AF503171.1 | - | 16 | 4869 | |||
AL833172.1 | - | 21 | 5354 | |||
BC112934.1 | - | 21 | 5343 | 6502888 | ||
BX648944.1 | - | 22 | 5364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
