siRNA Details
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324504.1 | NP_001311433.1 | 9 | 2494 | ||
NM_001324505.1 | NP_001311434.1 | 7 | 2462 | |||
NM_001324506.1 | NP_001311435.1 | 6 | 2368 | |||
NM_020382.4 | NP_065115.3 | 8 | 2584 | |||
NR_136910.1 | 9 | 2697 | ||||
XM_006719394.3 | XP_006719457.1 | 7 | 2414 | |||
XM_011538333.2 | XP_011536635.1 | 7 | 2800 | |||
GenBank mRNA | BC050346.1 | - | 9 | 2861 | 4838030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
