siRNA Details
Gene Symbol: SMURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199847.1 | NP_001186776.1 | 12, 13 | 1722 | ||
NM_020429.2 | NP_065162.1 | 13, 14 | 1800 | |||
NM_181349.2 | NP_851994.1 | 12, 13 | 1722 | |||
XM_017012457.1 | XP_016867946.1 | 13, 14 | 1798 | |||
GenBank mRNA | AB046845.1 | - | 13, 14 | 1768 | ||
AF199364.2 | - | 12, 13 | 1355 | |||
AF464850.1 | - | 13, 14 | 1460 | |||
AL834242.1 | - | 1, 2 | 83 | |||
BC136804.1 | - | 12, 13 | 1599 | 9020794 | ||
BC144414.1 | - | 12, 13 | 1599 | 9052935 | ||
BC152468.1 | - | 13, 14 | 1768 | 8862605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
