siRNA Details
Gene Symbol: USP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020935.2 | NP_065986.2 | 7, 8 | 1007 | ||
XM_005246720.2 | XP_005246777.1 | 7, 8 | 1079 | |||
XM_005246721.3 | XP_005246778.1 | 7, 8 | 971 | |||
XM_005246722.3 | XP_005246779.1 | 7, 8 | 1011 | |||
XM_005246724.1 | XP_005246781.1 | 7, 8 | 1007 | |||
XM_011511538.2 | XP_011509840.1 | 6, 7 | 952 | |||
XM_011511541.2 | XP_011509843.1 | 7, 8 | 1007 | |||
XM_017004577.1 | XP_016860066.1 | 7, 8 | 965 | |||
GenBank mRNA | AB046814.1 | - | 3, 4 | 452 | ||
AK095577.1 | - | 7, 8 | 965 | |||
AK128583.1 | - | 7, 8 | 1011 | |||
AL832645.2 | - | 7, 8 | 1006 | |||
BC050461.1 | - | 7, 8 | 976 | |||
BC112901.1 | - | 6, 7 | 807 | 6016503 | ||
BC133007.1 | - | 7, 8 | 892 | 40147029 | ||
BC133009.1 | - | 7, 8 | 937 | 40147031 | ||
BC144249.1 | - | 7, 8 | 892 | 9052769 | ||
BC144252.1 | - | 7, 8 | 932 | 9052772 | ||
BX538024.1 | - | 7, 8 | 972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
